Få mere at vide
N-[3-(Dimethylamino)propyl]lauramide Hydrochloride, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | N-[3-(Dimethylamino)propyl]lauramide Hydrochloride |
| CAS | 71732-95-1 |
| Molekylær formel | C17 H36 N2 O . H Cl |
| Mængde | 1 g |
| Synonym | N-[3-(Dimethylamino)propyl]dodecanamide Hydrochloride, (Laurylamidopropyl)dimethylamine Hydrochloride, 3-Dodecanamidopropyldimethylamine Hydrochloride, Dimethylaminopropyllaurylamide Hydrochloride, Lexamine L 13 Hydrochloride, Mackine 801 Hydrochloride, N,N-Dimethylaminopropyldodecylamide Hydrochloride, N-[3-(Dimethylamino)propyl]lauramide Hydrochloride, NSC 79892 Hydrochloride, N',N'-Dimethyl-N-dodecanoyl-1,3-diaminopropane Hydrochloride, Schercodine L Hydrochloride |
| InChI formel | InChI=1S/C17H36N2O.ClH/c1-4-5-6-7-8-9-10-11-12-14-17(20)18-15-13-16-19(2)3;/h4-16H2,1-3H3,(H,18,20);1H |
| SMIL | Cl.CCCCCCCCCCCC(=O)NCCCN(C)C |
| IUPAC navn | N-[3-(dimethylamino)propyl]dodecanamide;hydrochloride |
| Molekylvægt (g/mol) | 320.942 |
| Formel vægt | 320.259 |