Få mere at vide
N-(3-(Dimethylamino)propyl)dodecanamide, TRC
Artikelnummer. 30136916
Skift visning
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30136916 10 mg
30138556 100 mg
30008013 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30136916 Leverandør Toronto Research Chemicals Leverandørnr. TRCD45640510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 3179-80-4
Molekylær formel C17H36N2O
Molekylvægt (g/mol) 284.48
Synonym (Laurylamidopropyl)dimethylamine, 3-Dodecanamidopropyldimethylamine, 3-Lauramidopropyldimethylamine, Dimethylaminopropyllaurylamide, Lexamine L 13
IUPAC navn N-[3-(dimethylamino)propyl]dodecanamide
SMIL CCCCCCCCCCCC(=O)NCCCN(C)C
Tekniske data
Kemisk navn eller materiale N-(3-(Dimethylamino)propyl)dodecanamide
Mængde 10 mg
InChI formel InChI=1S/C17H36N2O/c1-4-5-6-7-8-9-10-11-12-14-17(20)18-15-13-16-19(2)3/h4-16H2,1-3H3,(H,18,20)
Formel vægt 284.2828