missing translation for 'onlineSavingsMsg'
Få mere at vide

N-(3-Chloro-4-((3-fluorobenzyl)oxy)phenyl)-6-iodoquinazolin-4-amine, 99%, BLD Pharm™

Artikelnummer. 30939036
Encompass_Preferred
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
250 mg
1 g
5 g
10 g
25 g
100 g
Pakningsstørrelse:
100g
10g
1g
250mg
25g
5g
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde Pakningsstørrelse
30939036 5 g 5g
30930758 10 g 10g
30934546 1 g 1g
30932469 25 g 25g
30931707 100 g 100g
30929743 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 30939036 Supplier BLD Pharm Supplier No. BD1425105g
 Begrænset vare
Forventet forsendelse: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Kemisk navn eller materiale N-(3-Chloro-4-((3-fluorobenzyl)oxy)phenyl)-6-iodoquinazolin-4-amine
CAS 231278-20-9
Molekylær formel C21H14ClFIN3O
MDL nummer MFCD09998827
Mængde 5 g
SMIL IC1=CC2=C(NC3=CC=C(OCC4=CC=CC(F)=C4)C(Cl)=C3)N=CN=C2C=C1
Molekylvægt (g/mol) 505.71
Procent renhed 99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.