Få mere at vide
N-(3-Carboxy-1-oxopropyl)glycylglycyl-N-(4-nitrophenyl)-L-phenylalaninamide, TRC
Artikelnummer. 30354175
Skift visning
Mængde:
10 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30354175 25 mg
30353460 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30354175 Leverandør Toronto Research Chemicals Leverandørnr. TRCC43286025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 68982-90-1
Molekylær formel C23H25N5O8
Molekylvægt (g/mol) 499.47
Synonym (S)-4-((2-((2-((1-((4-Nitrophenyl)amino)-1-oxo-3-phenylpropan-2-yl)amino)-2-oxoethyl)amino)-2-oxoethyl)amino)-4-oxobutanoic Acid, Chymotrypsin Substrate I, Colorimetric
IUPAC navn 4-[[2-[[2-[[(2S)-1-(4-nitroanilino)-1-oxo-3-phenylpropan-2-yl]amino]-2-oxoethyl]amino]-2-oxoethyl]amino]-4-oxobutanoic acid
SMIL OC(=O)CCC(=O)NCC(=O)NCC(=O)N[C@@H](Cc1ccccc1)C(=O)Nc2ccc(cc2)[N+](=O)[O-]
Tekniske data
Kemisk navn eller materiale N-(3-Carboxy-1-oxopropyl)glycylglycyl-N-(4-nitrophenyl)-L-phenylalaninamide
Mængde 25 mg
InChI formel InChI=1S/C23H25N5O8/c29-19(10-11-22(32)33)24-13-20(30)25-14-21(31)27-18(12-15-4-2-1-3-5-15)23(34)26-16-6-8-17(9-7-16)28(35)36/h1-9,18H,10-14H2,(H,24,29)(H,25,30)(H,26,34)(H,27,31)(H,32,33)/t18-/m0/s1
Formel vægt 499.1703