Få mere at vide
N-[3-Butanoyl-4-[(2RS)-2-hydroxy-3-[(1-methylethyl)amino]propoxy]phenyl]butanamide Hydrochloride, Mikromol™
Beskrivelse
N-[3-Butanoyl-4-[(2RS)-2-hydroxy-3-[(1-methylethyl)amino]propoxy]phenyl]butanamide Hydrochloride
Tekniske data
Tekniske data
| Kemisk navn eller materiale | N-[3-Butanoyl-4-[(2RS)-2-hydroxy-3-[(1-methylethyl)amino]propoxy]phenyl]butanamide Hydrochloride |
| CAS | 57898-71-2 |
| Analyt- eller komponentnavne | N-[3-Butanoyl-4-[(2RS)-2-hydroxy-3-[(1-methylethyl)amino]propoxy]phenyl]butanamide Hydrochloride |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C20 H32 N2 O4 . Cl H |
| Certificeringer/compliance | ISO 17025 |
| Synonym | N-[3-Butanoyl-4-[(2RS)-2-hydroxy-3-[(1-methylethyl)amino]propoxy]phenyl]butanamide Hydrochloride, Acebutolol Hydrochloride Imp. K (EP) as Hydrochloride, Acebutolol Hydrochloride Imp. K (EP), Acebutolol Imp. K (EP), Acebutolol Hydrochloride Impurity K as Hydrochloride, Acebutolol Impurity K as Hydrochloride |
| InChI formel | InChI=1S/C20H32N2O4.ClH/c1-5-7-18(24)17-11-15(22-20(25)8-6-2)9-10-19(17)26-13-16(23)12-21-14(3)4;/h9-11,14,16,21,23H,5-8,12-13H2,1-4H3,(H,22,25);1H |
| SMIL | Cl.CCCC(=O)Nc1ccc(OCC(O)CNC(C)C)c(c1)C(=O)CCC |
| IUPAC navn | N-[3-butanoyl-4-[2-hydroxy-3-(propan-2-ylamino)propoxy]phenyl]butanamide;hydrochloride |
| Vis mere |