Få mere at vide
N-[3-Acetyl-4-[(2RS)-2-hydroxy-3-[(1-methylethyl)amino]propoxy]phenyl]propanamide Hydrochloride, Mikromol
Beskrivelse
N-[3-Acetyl-4-[(2RS)-2-hydroxy-3-[(1-methylethyl)amino]propoxy]phenyl]propanamide Hydrochloride, Mikromol
Tekniske data
Tekniske data
| Kemisk navn eller materiale | N-[3-Acetyl-4-[(2RS)-2-hydroxy-3-[(1-methylethyl)amino]propoxy]phenyl]propanamide Hydrochloride |
| CAS | 57898-79-0 |
| Analyt- eller komponentnavne | N-[3-Acetyl-4-[(2RS)-2-hydroxy-3-[(1-methylethyl)amino]propoxy]phenyl]propanamide Hydrochloride |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C17 H26 N2 O4 . Cl H |
| Certificeringer/compliance | ISO 17025 |
| Synonym | N-[3-Acetyl-4-[(2RS)-2-hydroxy-3-[(1-methylethyl)amino]propoxy]phenyl]propanamide Hydrochloride, Acebutolol Hydrochloride Imp. J (EP) as Hydrochloride, Acebutolol Hydrochloride Imp. J (EP), Acebutolol Imp. J (EP), Acebutolol Hydrochloride Impurity J as Hydrochloride, Acebutolol Impurity J as Hydrochloride |
| InChI formel | InChI=1S/C17H26N2O4.ClH/c1-5-17(22)19-13-6-7-16(15(8-13)12(4)20)23-10-14(21)9-18-11(2)3;/h6-8,11,14,18,21H,5,9-10H2,1-4H3,(H,19,22);1H |
| Løsningstype | Neat |
| SMIL | Cl.CCC(=O)Nc1ccc(OCC(O)CNC(C)C)c(c1)C(=O)C |
| Vis mere |