Få mere at vide
N-[(3,4-Dimethoxy-2-pyridinyl)methyl] Pantoprazole Sulfone(Mixture of 1 and 3 isomers), TRC
Artikelnummer. 30379871
Skift visning
Mængde:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30379871 100 mg
30375004 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30379871 Leverandør Toronto Research Chemicals Leverandørnr. TRCD470970100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1797132-60-5
Molekylær formel C24H24F2N4O7S
Molekylvægt (g/mol) 550.53
Synonym 5-(Difluoromethoxy)-1-[(3,4-dimethoxy-2-pyridinyl)methyl]-2-[[(3,4-dimethoxy-2-pyridinyl)methyl]sulfonyl]-1H-benzimidazole, Pantoprazole Impurity
SMIL COc1ccnc(Cn2c(nc3cc(OC(F)F)ccc23)S(=O)(=O)Cc4nccc(OC)c4OC)c1OC
Tekniske data
Kemisk navn eller materiale N-[(3,4-Dimethoxy-2-pyridinyl)methyl] Pantoprazole Sulfone(Mixture of 1 and 3 isomers)
Mængde 100 mg
InChI formel InChI=1S/C24H24F2N4O7S/c1-33-19-7-9-27-16(21(19)35-3)12-30-18-6-5-14(37-23(25)26)11-15(18)29-24(30)38(31,32)13-17-22(36-4)20(34-2)8-10-28-17/h5-11,23H,12-13H2,1-4H3
Formel vægt 550.1334