Få mere at vide
N-[3-(4-Aminophenyl)propyl]-N,N-dimethylamine, TRC
Artikelnummer. 30327225
Skift visning
Mængde:
100 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30327225 250 mg
30323666 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30327225 Leverandør Toronto Research Chemicals Leverandørnr. TRCA615935250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-[3-(4-Aminophenyl)propyl]-N,N-dimethylamine
CAS 42817-60-7
Molekylær formel C11H18N2
Mængde 250 mg
Synonym 4-Amino-N,N-dimethylbenzenepropanamine, 4-(3-Dimethylaminopropyl)aniline, 4-(3-Dimethylaminopropyl)phenylamine, p-[3-(Dimethylamino)propyl]aniline
InChI formel InChI=1S/C11H18N2/c1-13(2)9-3-4-10-5-7-11(12)8-6-10/h5-8H,3-4,9,12H2,1-2H3
SMIL CN(C)CCCc1ccc(N)cc1
IUPAC navn 4-[3-(dimethylamino)propyl]aniline
Molekylvægt (g/mol) 178.27
Formel vægt 178.147