Få mere at vide
N-[3-(3-Chloro-10,11-dihydro-5H-dibenzo[b,f]azepin-5-yl)propyl]-N,N',N'-trimethylpropane-1,3-diamine Dihydrochloride, Mikromol™
Beskrivelse
N-[3-(3-Chloro-10,11-dihydro-5H-dibenzo[b,f]azepin-5-yl)propyl]-N,N',N'-trimethylpropane-1,3-diamine Dihydrochloride
Tekniske data
Tekniske data
| Kemisk navn eller materiale | N-[3-(3-Chloro-10,11-dihydro-5H-dibenzo[b,f]azepin-5-yl)propyl]-N,N',N'-trimethylpropane-1,3-diamine Dihydrochloride |
| CAS | 2305824-51-3 |
| Analyt- eller komponentnavne | N-[3-(3-Chloro-10,11-dihydro-5H-dibenzo[b,f]azepin-5-yl)propyl]-N,N',N'-trimethylpropane-1,3-diamine Dihydrochloride |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C23 H32 Cl N3 . 2 Cl H |
| Certificeringer/compliance | ISO 17025 |
| Synonym | Clomipramine Hydrochloride Imp. A (EP), Clomipramine Imp. A (EP), Clomipramine USP Related Compound A, Clomipramine USP RC A, N-[3-(3-Chloro-10,11-dihydro-5H-dibenzo[b,f]azepin-5-yl)propyl]-N,N',N'-trimethylpropane-1,3-diamine Dihydrochloride, Clomipramine Hydrochloride Imp. A (EP) as Dihydrochloride, Clomipramine Related Compound A as Dihydrochloride, Clomipramine Hydrochloride Impurity A as Dihydrochloride, Clomipramine Impurity A as Dihydrochloride |
| InChI formel | InChI=1S/C23H32ClN3.2ClH/c1-25(2)14-6-15-26(3)16-7-17-27-22-9-5-4-8-19(22)10-11-20-12-13-21(24)18-23(20)27;;/h4-5,8-9,12-13,18H,6-7,10-11,14-17H2,1-3H3;2*1H |
| SMIL | Cl.Cl.CN(C)CCCN(C)CCCN1c2ccccc2CCc3ccc(Cl)cc13 |
| IUPAC navn | N'-[3-(2-chloro-5,6-dihydrobenzo[b][1]benzazepin-11-yl)propyl]-N,N,N'-trimethylpropane-1,3-diamine;dihydrochloride |
| Vis mere |