Learn More
N-((2R,3S,4R,5S,6R)-2,4,5-Trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-3-yl)acetamide, 97%, BLD Pharm™
Specifications
Specifications
| CAS | 7772-94-3 |
| Molecular Formula | C8H15NO6 |
| MDL Number | MFCD00046758 |
| Quantity | 1 g |
| SMILES | CC(N[C@@H]1[C@H](O)O[C@H](CO)[C@@H](O)[C@@H]1O)=O |
| Molecular Weight (g/mol) | 221.21 |
| Percent Purity | 97% |
| Chemical Name or Material | N-((2R,3S,4R,5S,6R)-2,4,5-Trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-3-yl)acetamide |