Få mere at vide
N-[(2E)-3-(4-Hydroxyphenyl)-1-oxo-2-propen-1-yl]-L-aspartic Acid, TRC
Artikelnummer. 30179535
Skift visning
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30179535 50 mg
30179235 100 mg
30179432 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30179535 Leverandør Toronto Research Chemicals Leverandørnr. TRCH89998050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 151435-24-4
Molekylær formel C13H13NO6
Molekylvægt (g/mol) 279.25
Synonym (E)-N-[3-(4-Hydroxyphenyl)-1-oxo-2-propenyl]-L-aspartic Acid;
IUPAC navn (2S)-2-[[(E)-3-(4-hydroxyphenyl)prop-2-enoyl]amino]butanedioic acid
SMIL OC(=O)C[C@H](NC(=O)\C=C\c1ccc(O)cc1)C(=O)O
Tekniske data
Kemisk navn eller materiale N-[(2E)-3-(4-Hydroxyphenyl)-1-oxo-2-propen-1-yl]-L-aspartic Acid
InChI formel InChI=1S/C13H13NO6/c15-9-4-1-8(2-5-9)3-6-11(16)14-10(13(19)20)7-12(17)18/h1-6,10,15H,7H2,(H,14,16)(H,17,18)(H,19,20)/b6-3+/t10-/m0/s1
Mængde 50 mg
Formel vægt 279.0743