Få mere at vide
N-(2-Propenyl)-2-methylsulfonyl-10H-phenothiazine-d5, TRC
Artikelnummer. 30424624
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30424624 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30424624 Leverandør Toronto Research Chemicals Leverandørnr. TRCP76895225MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-(2-Propenyl)-2-methylsulfonyl-10H-phenothiazine-d5
CAS 1794940-43-4
Molekylær formel C16H10D5NO2S2
Mængde 25 mg
Synonym N-(2-Propenyl)-2-methylsulfonyl-phenothiazine-d5
InChI formel InChI=1S/C16H15NO2S2/c1-3-10-17-13-6-4-5-7-15(13)20-16-9-8-12(11-14(16)17)21(2,18)19/h3-9,11H,1,10H2,2H3/i1D2,3D,10D2
SMIL [2H]C(=C([2H])C([2H])([2H])N1c2ccccc2Sc3ccc(cc13)S(=O)(=O)C)[2H]
IUPAC navn 2-methylsulfonyl-10-(1,1,2,3,3-pentadeuterioprop-2-enyl)phenothiazine
Molekylvægt (g/mol) 322.46
Formel vægt 322.0858