Få mere at vide
N-(2-Nitrobenzyl)-N-cyclohexyl-N-methylamine, Mikromol™
Artikelnummer. 30052287
Skift visning
Mængde:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30052287 100 mg
30049996 25 mg
30462569 4 x 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30052287 Leverandør Mikromol Leverandørnr. MM0065.01
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
N-(2-Nitrobenzyl)-N-cyclohexyl-N-methylamine
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-(2-Nitrobenzyl)-N-cyclohexyl-N-methylamine
CAS 80638-08-0
Analyt- eller komponentnavne N-(2-Nitrobenzyl)-N-cyclohexyl-N-methylamine
Forsendelsestilstand Room Temperature
Molekylær formel C14 H20 N2 O2
Certificeringer/compliance ISO 17025
Synonym N-(2-Nitrobenzyl)-N-cyclohexyl-N-methylamine
InChI formel InChI=1S/C14H20N2O2/c1-15(13-8-3-2-4-9-13)11-12-7-5-6-10-14(12)16(17)18/h5-7,10,13H,2-4,8-9,11H2,1H3
SMIL CN(Cc1ccccc1[N+](=O)[O-])C2CCCCC2
IUPAC navn N-methyl-N-[(2-nitrophenyl)methyl]cyclohexanamine
Molekylvægt (g/mol) 248.32
Mængde 100 mg
Formel vægt 248.1525 g/mol