Få mere at vide
N-(2-Methylpropionyl-d7)glycine, CDN
Artikelnummer. 30321419
Skift visning
Mængde:
0.005 g
0.01 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30321419 0.005 g
30318390 0.01 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30321419 Leverandør CDN Leverandørnr. CDND76360.005G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-Isobutyrylglycine-D7 (isobutyryl-D7)
CAS 1330037-23-4
Molekylær formel C6H4D7NO3
Mængde 0.005 g
Synonym N-(2-methyl-1-oxopropyl-d7)glycine, N-Isobutyryl-d7-glycine;
InChI formel InChI=1 S/C6H11NO3/c1-4(2)6(10)7-3-5(8)9/h4H,3H2,1-2H3,(H,7,10)(H,8,9)/i1D3,2D3,4 D
SMIL O=C(C([2 H])(C([2 H])([2 H])[2 H])C([2 H])([2 H])[2 H])NCC(O)=O
IUPAC navn (2-(methyl-d3)propanoyl-2,3,3,3-d4)glycine
Molekylvægt (g/mol) 152.2
Formel vægt 152.12 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity