Få mere at vide
N-(2-Methylbenzoyl)glycine-2,2-d2, CDN
Artikelnummer. 30314303
Skift visning
Mængde:
0.1 g
0.25 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30314303 0.25 g -
30310126 0.1 g 0.1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30314303 Leverandør CDN Leverandørnr. CDND67980.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Methyl Hippuric Acid-d2
CAS 1185100-98-4
Molekylær formel C10 D2 H9 N O3
Mængde 0.25 g
Synonym 2,2-Dideuterio-2-[(2-methylbenzoyl)amino]acetic acid, N-(2-Methylbenzoyl)glycine-2,2-D2, N-(2-Methylbenzoyl)glycine-d2, 2-Methylhippuric acid-d2, 2-[(2-Methylbenzoyl)amino]acetic-2,2-d2 acid, o-Methylhippuric acid-d2, o-Toluric acid-d2, 2-Methyl hippuric acid-d2
InChI formel InChI=1 S/C10H11NO3/c1-7-4-2-3-5-8(7)10(14)11-6-9(12)13/h2-5 H,6H2,1H3,(H,11,14)(H,12,13)/i6D2
SMIL [2 H]C([2 H])(NC(=O)c1ccccc1C)C(=O)O
IUPAC navn 2,2-dideuterio-2-[(2-methylbenzoyl)amino]acetic acid
Molekylvægt (g/mol) 195.212
Formel vægt 195.086 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity