missing translation for 'onlineSavingsMsg'
Learn More

N-(2-Methyl-5-aminophenyl)guanidine, TRC

Product Code. 30476009
Change view
Click to view available options
Mængde:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30476009 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30476009 Supplier Toronto Research Chemicals Supplier No. TRCM692180500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale N-(2-Methyl-5-aminophenyl)guanidine
CAS 581076-64-4
Molekylær formel C8 H12 N4
Mængde 500 mg
Synonym Guanidine, N-(5-amino-2-methylphenyl)-, Guanidine, (5-amino-2-methylphenyl)- (9CI), N-(5-Amino-2-methylphenyl)guanidine, (5-Amino-2-methylphenyl)guanidine, N-(2-Methyl-5-aminophenyl)guanidine
InChI formel InChI=1S/C8H12N4/c1-5-2-3-6(9)4-7(5)12-8(10)11/h2-4H,9H2,1H3,(H4,10,11,12)
SMIL Cc1ccc(N)cc1NC(=N)N
IUPAC navn 1-(5-amino-2-methylphenyl)guanidine
Molekylvægt (g/mol) 164.208
Formel vægt 164.106
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.