missing translation for 'onlineSavingsMsg'
Learn More

N-(2-Methoxy-5-methylphenyl)benzenesulfonamide, 97%, Thermo Scientific™

Catalog Number 15492336
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
6964-02-9
MFCD00085470
This item is not returnable. View return policy
6964-02-9
MFCD00085470

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Molecular Formula C14H15NO3S
Quantity 250 mg
Synonym n-2-methoxy-5-methylphenyl benzenesulfonamide, 2'-methoxy-5'-methylbenzenesulfonanilide, benzenesulfonamide, n-2-methoxy-5-methylphenyl, 2-methoxy-5-methylphenyl phenylsulfonyl amine
InChI Key SNCSZUJMKFNFHX-UHFFFAOYSA-N
SMILES CC1=CC(=C(C=C1)OC)NS(=O)(=O)C2=CC=CC=C2
IUPAC Name N-(2-methoxy-5-methylphenyl)benzenesulfonamide
Molecular Weight (g/mol) 277.34
PubChem CID 248884
Formula Weight 277.34
Percent Purity 97%
Chemical Name or Material N-(2-Methoxy-5-methylphenyl)benzenesulfonamide
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'