Få mere at vide
N-2-Hydroxyethyl-N-(methyl-d3)-p-toluenesulfonamide, TRC
Artikelnummer. 30406062
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30406062 50 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30406062 Leverandør Toronto Research Chemicals Leverandørnr. TRCH94227250MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-2-Hydroxyethyl-N-(methyl-d3)-p-toluenesulfonamide
CAS 854634-39-2
Molekylær formel C10H12D3NO3S
Mængde 50 mg
InChI formel InChI=1S/C10H15NO3S/c1-9-3-5-10(6-4-9)15(13,14)11(2)7-8-12/h3-6,12H,7-8H2,1-2H3/i2D3
SMIL [2H]C([2H])([2H])N(CCO)S(=O)(=O)c1ccc(C)cc1
IUPAC navn N-(2-hydroxyethyl)-4-methyl-N-(trideuteriomethyl)benzenesulfonamide
Molekylvægt (g/mol) 232.31
Formel vægt 232.0961