Learn More
N-(2-Hydroxyethyl)-4-methyl-2-((4-methyl-1H-indol-3-yl)thio)pentamide, TRC
Product Code. 30404500
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Product Code. Quantity Unit Size
30404500 25 mg 25mg
1 options
Product Code. 30404500 Supplier Toronto Research Chemicals Supplier No. TRCH82558525MG
 Restricted Item
Forventet forsendelse: 10-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material N-(2-Hydroxyethyl)-4-methyl-2-((4-methyl-1H-indol-3-yl)thio)pentamide
CAS 1027997-01-8
Molecular Formula C17 H24 N2 O2 S
Quantity 25 mg
Synonym S 3969
InChI Formula InChI=1S/C17H24N2O2S/c1-11(2)9-14(17(21)18-7-8-20)22-15-10-19-13-6-4-5-12(3)16(13)15/h4-6,10-11,14,19-20H,7-9H2,1-3H3,(H,18,21)
SMILES CC(C)CC(Sc1c[nH]c2cccc(C)c12)C(=O)NCCO
IUPAC Name N-(2-hydroxyethyl)-4-methyl-2-[(4-methyl-1H-indol-3-yl)sulfanyl]pentanamide
Molecular Weight (g/mol) 320.45
Formula Weight 320.156