Få mere at vide
N-[2-Ethoxy-d5-1-[[2'-[(hydroxyamino)methyl][1,1'-biphenyl]-4-yl]methyl]-1H-benzimidazole-7-carboxylic Acid Methyl Ester]carbamic Acid 2-Ethylhexyl Ester, TRC
Artikelnummer. 30374194
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30374194 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30374194 Leverandør Toronto Research Chemicals Leverandørnr. TRCE8920371MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-(2-Ethoxy-d5-1-((2'-((hydroxyamino)methyl)(1,1'-biphenyl)-4-yl)methyl)-1H-benzimidazole-7-carboxylic Acid Methyl Ester)carbamic Acid 2-Ethylhexyl Ester
CAS 1795131-16-6
Molekylær formel C34H35D5N4O6
Mængde 1 mg
InChI formel InChI=1S/C34H40N4O6/c1-5-8-12-23(6-2)22-44-34(40)36-31(37-41)27-14-10-9-13-26(27)25-19-17-24(18-20-25)21-38-30-28(32(39)42-4)15-11-16-29(30)35-33(38)43-7-3/h9-11,13-20,23,41H,5-8,12,21-22H2,1-4H3,(H,36,37,40)/i3D3,7D2
SMIL [2H]C([2H])([2H])C([2H])([2H])Oc1nc2cccc(C(=O)OC)c2n1Cc3ccc(cc3)c4ccccc4C(=NC(=O)OCC(CC)CCCC)NO
IUPAC navn methyl 3-[[4-[2-[N'-(2-ethylhexoxycarbonyl)-N-hydroxycarbamimidoyl]phenyl]phenyl]methyl]-2-(1,1,2,2,2-pentadeuterioethoxy)benzimidazole-4-carboxylate
Molekylvægt (g/mol) 605.74
Formel vægt 605.3262