Få mere at vide
N-[2-(Diethylamino)ethyl]-5-formyl-2,4-dimethyl-1H-pyrrole-3-carboxamide, Mikromol
Artikelnummer. 30459725
Skift visning
Mængde:
25 mg
4 x 25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30459725 4 x 25 mg
30049959 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30459725 Leverandør Mikromol Leverandørnr. MM3568.0300254AMP
Discover Mikromol- your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
N-[2-(Diethylamino)ethyl]-5-formyl-2,4-dimethyl-1H-pyrrole-3-carboxamide, Mikromol
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-[2-(Diethylamino)ethyl]-5-formyl-2,4-dimethyl-1H-pyrrole-3-carboxamide
CAS 356068-86-5
Analyt- eller komponentnavne N-[2-(Diethylamino)ethyl]-5-formyl-2,4-dimethyl-1H-pyrrole-3-carboxamide
Forsendelsestilstand Room Temperature
Molekylær formel C14 H23 N3 O2
Certificeringer/compliance ISO 17025
Synonym N-[2-(Diethylamino)ethyl]-5-formyl-2,4-dimethyl-1H-pyrrole-3-carboxamide, 5-Formyl-2,4-dimethyl-1H-pyrrole-3-carboxylic acid (2-diethylaminoethyl)amide, 5-Formyl-2,4-dimethyl-1H-pyrrole-3-carboxylic acid N-(2-diethylaminoethyl)amide
InChI formel InChI=1S/C14H23N3O2/c1-5-17(6-2)8-7-15-14(19)13-10(3)12(9-18)16-11(13)4/h9,16H,5-8H2,1-4H3,(H,15,19)
Løsningstype Neat
SMIL CCN(CC)CCNC(=O)c1c(C)[nH]c(C=O)c1C
IUPAC navn N-[2-(diethylamino)ethyl]-5-formyl-2,4-dimethyl-1H-pyrrole-3-carboxamide
Molekylvægt (g/mol) 265.35
Mængde 4 x 25 mg
Formel vægt 265.179 g/mol