Få mere at vide
N-(2-Cyanoethyl-d3)-L-valine, TRC
Artikelnummer. 30360548
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
30360548 100 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30360548 Leverandør Toronto Research Chemicals Leverandørnr. TRCC981082100MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-(2-Cyanoethyl-(1,1,2-d3)) Valine
CAS 160210-22-0
Molekylær formel C8 2H3 H11 N2 O2
InChI formel InChI=1S/C8H14N2O2/c1-6(2)7(8(11)12)10-5-3-4-9/h6-7,10H,3,5H2,1-2H3,(H,11,12)/t7-/m0/s1/i3D,5D2/t3?,7-
SMIL [2H]C(C#N)C([2H])([2H])N[C@@H](C(C)C)C(=O)O
IUPAC navn (2S)-2-[(2-cyano-1,1,2-trideuterioethyl)amino]-3-methylbutanoic acid
Molekylvægt (g/mol) 173.23
Mængde 100 mg
Formel vægt 173.1244