missing translation for 'onlineSavingsMsg'
Få mere at vide

N-[(2'-Cyano[1,1'-biphenyl]-4-yl)methyl]-N-(1-oxopentyl)-L-valine Methyl Ester, TRC

Artikelnummer. 30360834
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30360834 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30360834 Leverandør Toronto Research Chemicals Leverandørnr. TRCC962370100MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale N-[(2'-Cyano[1,1'-biphenyl]-4-yl)methyl]-N-(1-oxopentyl)-L-valine Methyl Ester
CAS 137863-90-2
Molekylær formel C25 H30 N2 O3
Mængde 100 mg
Synonym L-Valine, N-[(2'-cyano[1,1'-biphenyl]-4-yl)methyl]-N-(1-oxopentyl)-, methyl ester, N-[(2'-Cyano[1,1'-biphenyl]-4-yl)methyl]-N-(1-oxopentyl)-L-valine methyl ester
InChI formel InChI=1S/C25H30N2O3/c1-5-6-11-23(28)27(24(18(2)3)25(29)30-4)17-19-12-14-20(15-13-19)22-10-8-7-9-21(22)16-26/h7-10,12-15,18,24H,5-6,11,17H2,1-4H3/t24-/m0/s1
SMIL CCCCC(=O)N(Cc1ccc(cc1)c2ccccc2C#N)[C@@H](C(C)C)C(=O)OC
IUPAC navn methyl (2S)-2-[[4-(2-cyanophenyl)phenyl]methyl-pentanoylamino]-3-methylbutanoate
Molekylvægt (g/mol) 406.52
Formel vægt 406.2256
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.