Få mere at vide
N-[2-Chloro-4-(trifluoromethyl)phenyl]-D-valine, TRC
Artikelnummer. 30137549
Skift visning
Mængde:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30137549 1 g
30009212 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30137549 Leverandør Toronto Research Chemicals Leverandørnr. TRCC4217851G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 76769-07-8
Molekylær formel C12H13ClF3NO2
Molekylvægt (g/mol) 295.69
Synonym (R)-2-((2-Chloro-4-(trifluoromethyl)phenyl)amino)-3-methylbutanoic Acid
IUPAC navn (2R)-2-[2-chloro-4-(trifluoromethyl)anilino]-3-methylbutanoic acid
SMIL CC(C)[C@@H](Nc1ccc(cc1Cl)C(F)(F)F)C(=O)O
Tekniske data
Kemisk navn eller materiale N-[2-Chloro-4-(trifluoromethyl)phenyl]-D-valine
InChI formel InChI=1S/C12H13ClF3NO2/c1-6(2)10(11(18)19)17-9-4-3-7(5-8(9)13)12(14,15)16/h3-6,10,17H,1-2H3,(H,18,19)/t10-/m1/s1
Mængde 1 g
Formel vægt 295.0587