missing translation for 'onlineSavingsMsg'
Learn More

N-(2-Bromo-5-nitrophenyl)benzenesulfonamide, 97%, Thermo Scientific™

Catalog Number p-7037468
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
1421261-66-6
C12H9BrN2O4S
357.18
MFCD22682850
KTKONBLIZGUGBA-UHFFFAOYSA-N
n-2-bromo-5-nitrophenyl benzenesulfonamide
73996217
N-(2-bromo-5-nitrophenyl)benzenesulfonamide
[O-][N+](=O)C1=CC(NS(=O)(=O)C2=CC=CC=C2)=C(Br)C=C1
This item is not returnable. View return policy
1421261-66-6
C12H9BrN2O4S
357.18
MFCD22682850
KTKONBLIZGUGBA-UHFFFAOYSA-N
n-2-bromo-5-nitrophenyl benzenesulfonamide
73996217
N-(2-bromo-5-nitrophenyl)benzenesulfonamide
[O-][N+](=O)C1=CC(NS(=O)(=O)C2=CC=CC=C2)=C(Br)C=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 357.18
Percent Purity 97%
Chemical Name or Material N-(2-Bromo-5-nitrophenyl)benzenesulfonamide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'