missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(2-Bromo-4-methylphenyl)-2,2-diphenylacetamide, 95%

Brand:  Alfa Aesar™ H65902.03

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15469986

  • 2009.39 DKK / 1g


Estimated Shipment date
from Supplier 17-10-2018
Add to basket

Specifications

Chemical Identifiers

MFCD03384050

Specifications

N-(2-Bromo-4-methylphenyl)-2,2-diphenylacetamide
MFCD03384050
380.29
95%
1g
Solid
Videos
SDS