missing translation for 'onlineSavingsMsg'
Få mere at vide

N-(2-Aminoethyl)-2-(naphthalen-1-yl)acetamide Hydrochloride (Naphthylacetylethylenediamine Hydrochloride), Mikromol™

Artikelnummer. 17961736
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17961736 100 mg
17904376 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 17961736 Leverandør Mikromol Leverandørnr. MM0075.06

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

N-(2-Aminoethyl)-2-(naphthalen-1-yl)acetamide Hydrochloride (Naphthylacetylethylenediamine Hydrochloride)
TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale N-(2-Aminoethyl)-2-(naphthalen-1-yl)acetamide Hydrochloride (Naphthylacetylethylenediamine Hydrochloride)
CAS 73664-32-1
Analyt- eller komponentnavne N-(2-Aminoethyl)-2-(naphthalen-1-yl)acetamide Hydrochloride (Naphthylacetylethylenediamine Hydrochloride)
Forsendelsestilstand Room Temperature
Molekylær formel C14 H16 N2 O . Cl H
Certificeringer/compliance ISO 9001
Synonym Naphazoline Hydrochloride Imp. A (EP), Naphazoline Imp. A (EP), Naphazoline USP Related Compound A, Naphazoline USP RC A, N-(2-Aminoethyl)-2-(naphthalen-1-yl)acetamide Hydrochloride, Naphazoline Nitrate Imp. A (EP) as Hydrochloride, Naphazoline Hydrochloride Imp. A (EP) as Hydrochloride, Naphthylacetylethylenediamine Hydrochloride, Naphazoline Related Compound A as Hydrochloride, Naphazoline Hydrochloride Impurity A as Hydrochloride, Naphazoline Nitrate Impurity A as Hydrochloride, Naphazoline Impurity A as Hydrochloride, Naphazoline Impurity A as Hydrochloride
InChI formel InChI=1S/C14H16N2O.ClH/c15-8-9-16-14(17)10-12-6-3-5-11-4-1-2-7-13(11)12;/h1-7H,8-10,15H2,(H,16,17);1H
SMIL Cl.NCCNC(=O)Cc1cccc2ccccc12
IUPAC navn N-(2-aminoethyl)-2-naphthalen-1-ylacetamide;hydrochloride
Molekylvægt (g/mol) 264.75
Mængde 100 mg
Formel vægt 264.1029 g/mol
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.