Få mere at vide
N-(2-Aminoethyl)-1,2,3,4-tetrahydronaphthalene-1-yl-carboxamide Hydrochloride ((1,2,3,4-Tetrahydro-1-naphthyl)formylethylenediamine Hydrochloride), Mikromol™
Artikelnummer. 17973496
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17973496 100 mg
17954306 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 17973496 Leverandør Mikromol Leverandørnr. MM0088.05
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
N-(2-Aminoethyl)-1,2,3,4-tetrahydronaphthalene-1-yl-carboxamide Hydrochloride ((1,2,3,4-Tetrahydro-1-naphthyl)formylethylenediamine Hydrochloride)
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-(2-Aminoethyl)-1,2,3,4-tetrahydronaphthalene-1-yl-carboxamide hydrochloride
CAS 1828966-95-5
Analyt- eller komponentnavne N-(2-Aminoethyl)-1,2,3,4-tetrahydronaphthalene-1-yl-carboxamide hydrochloride
Forsendelsestilstand Room Temperature
Molekylær formel C13 H18 N2 O . Cl H
Certificeringer/compliance ISO 17025
InChI formel InChI=1S/C13H18N2O.ClH/c14-8-9-15-13(16)12-7-3-5-10-4-1-2-6-11(10)12;/h1-2,4,6,12H,3,5,7-9,14H2,(H,15,16);1H
IUPAC navn N-(2-aminoethyl)-1,2,3,4-tetrahydronaphthalene-1-carboxamide;hydrochloride
Molekylvægt (g/mol) 254.7558