Få mere at vide
N-(2,6-Dimethylphenyl)-2-[(1-methylethyl)amino]acetamide Hydrochloride, Mikromol™
Artikelnummer. 30458665
Skift visning
Mængde:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30458665 4 x 25 mg
30051404 25 mg
17900746 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30458665 Leverandør Mikromol Leverandørnr. MM0102.1100254AMP
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
N-(2,6-Dimethylphenyl)-2-[(1-methylethyl)amino]acetamide Hydrochloride, Mikromol
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-(2,6-Dimethylphenyl)-2-[(1-methylethyl)amino]acetamide Hydrochloride
CAS 35891-87-3
Analyt- eller komponentnavne N-(2,6-Dimethylphenyl)-2-[(1-methylethyl)amino]acetamide Hydrochloride
Forsendelsestilstand Room Temperature
Molekylær formel C13 H20 N2 O . Cl H
Certificeringer/compliance ISO 17025
Synonym Lidocaine Hydrochloride Imp. G (EP), Lidocaine Imp. G (EP), N-(2,6-Dimethylphenyl)-2-[(1-methylethyl)amino]acetamide Hydrochloride, Lidocaine Hydrochloride Imp. G (EP) as Hydrochloride, Lidocaine Hydrochloride Impurity G as Hydrochloride, Lidocaine Impurity G as Hydrochloride
InChI formel InChI=1S/C13H20N2O.ClH/c1-9(2)14-8-12(16)15-13-10(3)6-5-7-11(13)4;/h5-7,9,14H,8H2,1-4H3,(H,15,16);1H
Løsningstype Neat
SMIL Cl.CC(C)NCC(=O)Nc1c(C)cccc1C
IUPAC navn N-(2,6-dimethylphenyl)-2-(propan-2-ylamino)acetamide;hydrochloride
Molekylvægt (g/mol) 256.77
Mængde 4 x 25 mg
Formel vægt 256.1342 g/mol