missing translation for 'onlineSavingsMsg'
Learn More

N-(2,6-Dibromo-3-pyridyl)thiourea, 97+%, Thermo Scientific™

Catalog Number 15446517
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
1355241-48-3
C6H5Br2N3S
311.00
MFCD22682809
JYUVZMXPLAAKLL-UHFFFAOYSA-N
1-2,6-dibromopyridin-3-yl thiourea, n-2,6-dibromo-3-pyridyl thiourea
73995160
(2,6-dibromopyridin-3-yl)thiourea
NC(=S)NC1=C(Br)N=C(Br)C=C1
This item is not returnable. View return policy
1355241-48-3
C6H5Br2N3S
311.00
MFCD22682809
JYUVZMXPLAAKLL-UHFFFAOYSA-N
1-2,6-dibromopyridin-3-yl thiourea, n-2,6-dibromo-3-pyridyl thiourea
73995160
(2,6-dibromopyridin-3-yl)thiourea
NC(=S)NC1=C(Br)N=C(Br)C=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 228°C to 232°C
Quantity 1 g
Formula Weight 310.99
Percent Purity ≥97%
Chemical Name or Material N-(2,6-Dibromo-3-pyridyl)thiourea
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'