Få mere at vide
N-[2(4-(-Octylphenyl))butanoic Acid] Fingolimod, TRC
Artikelnummer. 30420355
Skift visning
Mængde:
0.5 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30420355 10 mg
30410593 0.5 mg
30427406 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30420355 Leverandør Toronto Research Chemicals Leverandørnr. TRCO29385510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-[2(4-(-Octylphenyl))butanoic Acid] Fingolimod
CAS 2147726-86-9
Analyt- eller komponentnavne N-[2(4-(-Octylphenyl))butanoic Acid] Fingolimod
Forsendelsestilstand Room Temperature
Molekylær formel C37H59NO4
Synonym 2-((1-Hydroxy-2-(hydroxymethyl)-4-(4-octylphenyl)butan-2-yl)amino)-4-(4-octylphenyl)butanoic Acid
InChI formel InChI=1S/C37H59NO4/c1-3-5-7-9-11-13-15-31-17-21-33(22-18-31)25-26-35(36(41)42)38-37(29-39,30-40)28-27-34-23-19-32(20-24-34)16-14-12-10-8-6-4-2/h17-24,35,38-40H,3-16,25-30H2,1-2H3,(H,41,42)
Løsningstype Neat
SMIL OC(C(NC(CCC1=CC=C(CCCCCCCC)C=C1)(CO)CO)CCC2=CC=C(CCCCCCCC)C=C2)=O
Industry Type Pharmaceutical
Molekylvægt (g/mol) 581.87