missing translation for 'onlineSavingsMsg'
Få mere at vide

N-[2-(4-Methoxyphenyl)-1-methylethyl]-formamide, TRC

Artikelnummer. 30468072
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30468072 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30468072 Leverandør Toronto Research Chemicals Leverandørnr. TRCM265885100MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kræver en prisopdatering, inden den kan bestilles.

Anmod om en pris på denne vare her.

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale N-[2-(4-Methoxyphenyl)-1-methylethyl]-formamide
CAS 126002-14-0
Molekylær formel C11H15NO2
Mængde 100 mg
Synonym Formoterol Impurity 23
InChI formel InChI=1S/C11H15NO2/c1-9(12-8-13)7-10-3-5-11(14-2)6-4-10/h3-6,8-9H,7H2,1-2H3,(H,12,13)
SMIL O=CNC(CC1=CC=C(C=C1)OC)C
IUPAC navn N-(1-(4-methoxyphenyl)propan-2-yl)formamide
Molekylvægt (g/mol) 193.24
Formel vægt 193.11
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.