Få mere at vide
N-2,3-Xylyl-N-nitrosoanthranilic Acid, TRC
Artikelnummer. 30137675
Skift visning
Mængde:
50 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30137675 500 mg
30134567 250 mg
30009265 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30137675 Leverandør Toronto Research Chemicals Leverandørnr. TRCN481270500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 2114-63-8
Molekylær formel C15 H14 N2 O3
Molekylvægt (g/mol) 270.283
Synonym 2-[(2,3-Dimethylphenyl)nitrosoamino]benzoic Acid, ITF 616;
IUPAC navn 2-(2,3-dimethyl-N-nitrosoanilino)benzoic acid
SMIL Cc1cccc(N(N=O)c2ccccc2C(=O)O)c1C
Tekniske data
Kemisk navn eller materiale N-2,3-Xylyl-N-nitrosoanthranilic Acid
Mængde 500 mg
InChI formel InChI=1S/C15H14N2O3/c1-10-6-5-9-13(11(10)2)17(16-20)14-8-4-3-7-12(14)15(18)19/h3-9H,1-2H3,(H,18,19)
Formel vægt 270.1