Få mere at vide
N-(2,3-Dimethylphenyl) Mefenamic Acid Carboxamide, TRC
Artikelnummer. 30378556
Skift visning
Mængde:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30378556 10 g
30377674 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30378556 Leverandør Toronto Research Chemicals Leverandørnr. TRCD47794510G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 21122-68-9
Molekylær formel C23 H24 N2 O
Molekylvægt (g/mol) 344.45
Synonym N-(2,3-Dimethylphenyl)-2-[(2,3-dimethylphenyl)amino]benzamide, Mefenamic Acid Imp. B (EP)
IUPAC navn 2-(2,3-dimethylanilino)-N-(2,3-dimethylphenyl)benzamide
SMIL Cc1cccc(NC(=O)c2ccccc2Nc3cccc(C)c3C)c1C
Tekniske data
Kemisk navn eller materiale N-(2,3-Dimethylphenyl)-2-[(2,3-dimethylphenyl)amino]benzamide
Mængde 10 g
InChI formel InChI=1S/C23H24N2O/c1-15-9-7-13-20(17(15)3)24-22-12-6-5-11-19(22)23(26)25-21-14-8-10-16(2)18(21)4/h5-14,24H,1-4H3,(H,25,26)
Formel vægt 344.1889