Få mere at vide
N-(2,2,2-Trichloroethoxy)carbonyl] Nortilidine-d3, TRC
Artikelnummer. 30449376
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30449376 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30449376 Leverandør Toronto Research Chemicals Leverandørnr. TRCT7741721MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-[(2,2,2-Trichloroethoxy)carbonyl] Nortilidine-d3
CAS 1246820-72-3
Molekylær formel C19H19D3Cl3NO4
Mængde 1 mg
Synonym trans-1-phenyl-N-(methyl-d3)-2-[[(2,2,2-trichloroethoxy)carbonyl]amino]-3-cyclohexene-1-carboxylic Acid Ethyl Ester
InChI formel InChI=1S/C19H22Cl3NO4/c1-3-26-16(24)18(14-9-5-4-6-10-14)12-8-7-11-15(18)23(2)17(25)27-13-19(20,21)22/h4-7,9-11,15H,3,8,12-13H2,1-2H3/t15-,18-/m1/s1/i2D3
SMIL [2H]C([2H])([2H])N([C@@H]1C=CCC[C@@]1(C(=O)OCC)c2ccccc2)C(=O)OCC(Cl)(Cl)Cl
IUPAC navn ethyl (1R,2R)-1-phenyl-2-[2,2,2-trichloroethoxycarbonyl(trideuteriomethyl)amino]cyclohex-3-ene-1-carboxylate
Molekylvægt (g/mol) 437.76
Formel vægt 436.0803