Learn More
N-[(1R)-1-(4-Acetylphenyl)ethyl]acetamide, TRC
Product Code. 30327620
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
Product Code. Quantity Unit Size
30327620 500 mg 500mg
1 options
Product Code. 30327620 Supplier Toronto Research Chemicals Supplier No. TRCA187385500MG
 Restricted Item
Forventet forsendelse: 10-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material N-[(1R)-1-(4-Acetylphenyl)ethyl]acetamide
CAS 88146-37-6
Molecular Formula C12H15NO2
Quantity 500 mg
Synonym (+)-N-[1-(4-Acetylphenyl)ethyl]acetamide
InChI Formula InChI=1S/C12H15NO2/c1-8(13-10(3)15)11-4-6-12(7-5-11)9(2)14/h4-8H,1-3H3,(H,13,15)/t8-/m1/s1
SMILES C[C@@H](NC(=O)C)c1ccc(cc1)C(=O)C
IUPAC Name N-[(1R)-1-(4-acetylphenyl)ethyl]acetamide
Molecular Weight (g/mol) 205.25
Formula Weight 205.1103