missing translation for 'onlineSavingsMsg'
Learn More

N-(1-Naphthyl)thiourea, 97%, Thermo Scientific™

Catalog Number p-7024412
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
86-88-4
C11H10N2S
202.275
MFCD00041824
PIVQQUNOTICCSA-UHFFFAOYSA-N
1-1-naphthyl-2-thiourea, antu, 1-naphthylthiourea, dirax, anturat, rattrack, smeesana, alrato, bantu, kripid
736366
CHEBI:82256
naphthalen-1-ylthiourea
C1=CC=C2C(=C1)C=CC=C2NC(=S)N
This item is not returnable. View return policy
86-88-4
C11H10N2S
202.275
MFCD00041824
PIVQQUNOTICCSA-UHFFFAOYSA-N
1-1-naphthyl-2-thiourea, antu, 1-naphthylthiourea, dirax, anturat, rattrack, smeesana, alrato, bantu, kripid
736366
CHEBI:82256
naphthalen-1-ylthiourea
C1=CC=C2C(=C1)C=CC=C2NC(=S)N

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 185°C to 191°C
Odor Odorless
Quantity 10 g
UN Number UN1651
Beilstein 778118
Merck Index 14,722
Formula Weight 202.29
Percent Purity 97%
Chemical Name or Material N-(1-Naphthyl)thiourea
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'