Få mere at vide
N-(1-Naphthyl-d7-methyl)methylamine, TRC
Artikelnummer. 30483572
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
30483572 100 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30483572 Leverandør Toronto Research Chemicals Leverandørnr. TRCN372952100MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-(1-Naphthyl-d7-methyl)methylamine
CAS 1189686-07-4
Molekylær formel C12 2H7 H6 N
Mængde 100 mg
InChI formel InChI=1S/C12H13N/c1-13-9-11-7-4-6-10-5-2-3-8-12(10)11/h2-8,13H,9H2,1H3/i2D,3D,4D,5D,6D,7D,8D
SMIL [2H]c1c([2H])c([2H])c2c(CNC)c([2H])c([2H])c([2H])c2c1[2H]
IUPAC navn 1-(2,3,4,5,6,7,8-heptadeuterionaphthalen-1-yl)-N-methylmethanamine
Molekylvægt (g/mol) 178.28
Formel vægt 178.1487