Få mere at vide
N-[1-(3’-Benzyloxyphenyl)ethyl]-N,N-dimethyl-d6-amine, TRC
Artikelnummer. 30343679
Skift visning
Mængde:
2.5 mg
25 mg
Pakningsstørrelse:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30343679 25 mg -
30337017 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30343679 Leverandør Toronto Research Chemicals Leverandørnr. TRCB28768725MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-[1-(3'-Benzyloxyphenyl)ethyl]-N,N-dimethyl-d6-amine
CAS 1190006-94-0
Molekylær formel C17 H15 D6 N O
Mængde 25 mg
InChI formel InChI=1S/C17H21NO/c1-14(18(2)3)16-10-7-11-17(12-16)19-13-15-8-5-4-6-9-15/h4-12,14H,13H2,1-3H3/i2D3,3D3
SMIL [2H]C([2H])([2H])N(C(C)c1cccc(OCc2ccccc2)c1)C([2H])([2H])[2H]
IUPAC navn 1-(3-phenylmethoxyphenyl)-N,N-bis(trideuteriomethyl)ethanamine
Molekylvægt (g/mol) 261.39
Formel vægt 261.2