Få mere at vide
N-[1-(3’-Benzyloxyphenyl)ethyl]-N-(methyl-d3)-O-ethylcarbamate, TRC
Artikelnummer. 30347152
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30347152 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30347152 Leverandør Toronto Research Chemicals Leverandørnr. TRCB2876975MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-[1-(3'-Benzyloxyphenyl)ethyl]-N-(methyl-d3)-O-ethylcarbamate
CAS 1189709-65-6
Molekylær formel C19 H20 D3 N O3
Mængde 5 mg
InChI formel InChI=1S/C19H23NO3/c1-4-22-19(21)20(3)15(2)17-11-8-12-18(13-17)23-14-16-9-6-5-7-10-16/h5-13,15H,4,14H2,1-3H3/i3D3
SMIL [2H]C([2H])([2H])N(C(C)c1cccc(OCc2ccccc2)c1)C(=O)OCC
IUPAC navn ethyl N-[1-(3-phenylmethoxyphenyl)ethyl]-N-(trideuteriomethyl)carbamate
Molekylvægt (g/mol) 316.41
Formel vægt 316.1866