Få mere at vide
N-[1-(3’-Benzyloxyphenyl)ethyl]-N-(methyl-d3)amine, TRC
Artikelnummer. 30339709
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
30339709 100 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30339709 Leverandør Toronto Research Chemicals Leverandørnr. TRCB287692100MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-[1-(3'-Benzyloxyphenyl)ethyl]-N-(methyl-d3)amine
CAS 1189957-44-5
Molekylær formel C16H16D3NO
Mængde 100 mg
InChI formel InChI=1S/C16H19NO/c1-13(17-2)15-9-6-10-16(11-15)18-12-14-7-4-3-5-8-14/h3-11,13,17H,12H2,1-2H3/i2D3
SMIL [2H]C([2H])([2H])NC(C)c1cccc(OCc2ccccc2)c1
IUPAC navn 1-(3-phenylmethoxyphenyl)-N-(trideuteriomethyl)ethanamine
Molekylvægt (g/mol) 244.35
Formel vægt 244.1655