Få mere at vide
N-(1,3,4-Thiadiazolyl)nicotinamide, TRC
Artikelnummer. 17944815
Skift visning
Mængde:
5 mg
25 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17944815 25 mg
17912105 1 g
17981555 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 17944815 Leverandør Toronto Research Chemicals Leverandørnr. TRCT34400025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 51987-99-6
Molekylær formel C8H6N4OS
Molekylvægt (g/mol) 206.22
Synonym N-(1,3,4-Thiadiazol-2-yl)pyridine-3-carboxamide, N-(1,3,4-Thiadiazolyl)-m-nicotinamide;
SMIL O=C(NC1=NN=CS1)C2=CC=CN=C2
Tekniske data
Kemisk navn eller materiale N-(1,3,4-Thiadiazolyl)nicotinamide
Renhedsgrad noter HPLC
Mængde 25 mg
InChI formel InChI=1S/C8H6N4OS/c13-7(6-2-1-3-9-4-6)11-8-12-10-5-14-8/h1-5H,(H,11,12,13)
Formel vægt 206.0262
Procent renhed >95%