Få mere at vide
N-((1-(2-(tert-Butylamino)-2-oxoethyl)piperidin-4-yl)methyl)-3-chloro-5-fluorobenzamide-d9 Hydrochlo, TRC
Artikelnummer. 30340479
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30340479 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30340479 Leverandør Toronto Research Chemicals Leverandørnr. TRCB6906671MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-((1-(2-(tert-Butylamino)-2-oxoethyl)piperidin-4-yl)methyl)-3-chloro-5-fluorobenzamide-d9 Hydrochloride Salt
CAS 1346604-21-4
Molekylær formel C19H19D9Cl2FN3O2
Mængde 1 mg
Synonym 4-[[(3-Chloro-5-fluorobenzoyl)amino]methyl]-N-(1,1-dimethylethyl-d9)-1-piperidineacetamide Hydrochloride Salt
InChI formel InChI=1S/C19H27ClFN3O2.ClH/c1-19(2,3)23-17(25)12-24-6-4-13(5-7-24)11-22-18(26)14-8-15(20)10-16(21)9-14;/h8-10,13H,4-7,11-12H2,1-3H3,(H,22,26)(H,23,25);1H/i1D3,2D3,3D3;
SMIL ClC1=CC(F)=CC(C(NCC2CCN(CC(NC(C([2H])([2H])[2H])(C([2H])([2H])[2H])C([2H])([2H])[2H])=O)CC2)=O)=C1.Cl
IUPAC navn 3-chloro-5-fluoro-N-[[1-[2-[[1,1,1,3,3,3-hexadeuterio-2-(trideuteriomethyl)propan-2-yl]amino]-2-oxoethyl]piperidin-4-yl]methyl]benzamide;hydrochloride
Molekylvægt (g/mol) 429.4