Få mere at vide
N-[(1,1-Dimethylethoxy)carbonyl]glycine-2,2-d2, TRC
Artikelnummer. 30379204
Skift visning
Mængde:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30379204 10 mg
30376251 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30379204 Leverandør Toronto Research Chemicals Leverandørnr. TRCD47292810MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-[(1,1-Dimethylethoxy)carbonyl]glycine-2,2-d2
CAS 42492-65-9
Molekylær formel C7 D2 H11 N O4
Mængde 10 mg
Synonym Glycine-2,2-d2, N-[(1,1-dimethylethoxy)carbonyl]- (9CI, ACI), N-[(1,1-Dimethylethoxy)carbonyl]glycine-2,2-d2 (ACI), 2-(tert-Butoxycarbonylamino)-2,2-dideuterio-acetic acid, N-(tert-Butoxycarbonyl)glycine-d2, N-(tert-Butoxycarbonyl)glycine-2,2-D2, N-Boc-glycine-2,2-d2, Glycine-2,2-d2-N-t-BOC
InChI formel InChI=1S/C7H13NO4/c1-7(2,3)12-6(11)8-4-5(9)10/h4H2,1-3H3,(H,8,11)(H,9,10)/i4D2
SMIL [2H]C([2H])(NC(=O)OC(C)(C)C)C(=O)O
IUPAC navn 2,2-dideuterio-2-[(2-methylpropan-2-yl)oxycarbonylamino]acetic acid
Molekylvægt (g/mol) 177.195
Formel vægt 177.097