Få mere at vide
(-)-Myrtenol, TRC
Artikelnummer. 30005736
Skift visning
Mængde:
1 g
5 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30005736 5 g
30010136 10 g
30006451 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30005736 Leverandør Toronto Research Chemicals Leverandørnr. TRCM8850635G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 19894-97-4
Molekylær formel C10 H16 O
Molekylvægt (g/mol) 152.23
Synonym (1R,5S)-6,6-Dimethyl-bicyclo(3.1.1)hept-2-ene-2-methanol
IUPAC navn [(1R,5S)-6,6-dimethyl-4-bicyclo[3.1.1]hept-3-enyl]methanol
SMIL CC1(C)[C@@H]2CC=C(CO)[C@H]1C2
Tekniske data
Kemisk navn eller materiale (-)-Myrtenol
Mængde 5 g
InChI formel InChI=1S/C10H16O/c1-10(2)8-4-3-7(6-11)9(10)5-8/h3,8-9,11H,4-6H2,1-2H3/t8-,9-/m1/s1
Formel vægt 152.1201