Få mere at vide
Myristoylamide Propylbetaine, TRC
Artikelnummer. 30134810
Skift visning
Mængde:
250 mg
2.5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30134810 2.5 g
30009906 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30134810 Leverandør Toronto Research Chemicals Leverandørnr. TRCM8847352.5G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 59272-84-3
Molekylær formel C21 H42 N2 O3
Molekylvægt (g/mol) 370.5698
Synonym 1-Propanaminium, N-(carboxymethyl)-N,N-dimethyl-3-[(1-oxotetradecyl)amino]-, hydroxide, inner salt, ACB 14, Marpobistar MAP, Myristamidopropyl betaine, Myristylamidopropylbetaine, Softazoline MPB, Vista MAP
IUPAC navn 2-[dimethyl-[3-(tetradecanoylamino)propyl]azaniumyl]acetate
SMIL CCCCCCCCCCCCCC(=O)NCCC[N+](C)(C)CC(=O)[O-]
Tekniske data
Kemisk navn eller materiale Myristamidopropyl Betaine
Mængde 2.5 g
InChI formel InChI=1S/C21H42N2O3/c1-4-5-6-7-8-9-10-11-12-13-14-16-20(24)22-17-15-18-23(2,3)19-21(25)26/h4-19H2,1-3H3,(H-,22,24,25,26)
Formel vægt 370.3195