Få mere at vide
Myristoyl-L-carnitine-d3 Hydrochloride, TRC
Artikelnummer. 30470345
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30470345 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30470345 Leverandør Toronto Research Chemicals Leverandørnr. TRCM88474725MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Myristoyl-L-carnitine-d3 Hydrochloride
CAS 1334532-25-0
Molekylær formel C21 D3 H39 N O4 . Cl
Mængde 25 mg
Synonym (R)-3-Carboxy-N,N,N-trimethyl-2-[(1-oxotetradecyl)oxy]-1-propanaminium-d3 Chloride, Tetradecanoyl-L-carnitine-(N-methyl-d3) Hydrochloride, (3R)-4-(Dimethyl-(trideuteriomethyl)azaniumyl)-3-tetradecanoyloxybutanoate hydrochloride, Tetradecanoyl-L-carnitine-d3 HCl (N-methyl-d3), Myristoyl-L-carnitine-D3 (N-methyl-D3) chloride, (2R)-3-Carboxy-N,N-dimethyl-N-(methyl-d3)-2-[(1-oxotetradecyl)oxy]-1-propanaminium chloride
InChI formel InChI=1S/C21H41NO4.ClH/c1-5-6-7-8-9-10-11-12-13-14-15-16-21(25)26-19(17-20(23)24)18-22(2,3)4;/h19H,5-18H2,1-4H3;1H/t19-;/m1./s1/i2D3;
SMIL [Cl-].[2H]C([2H])([2H])[N+](C)(C)C[C@@H](CC(=O)O)OC(=O)CCCCCCCCCCCCC
IUPAC navn [(2R)-3-carboxy-2-tetradecanoyloxypropyl]-dimethyl-(trideuteriomethyl)azanium;chloride
Molekylvægt (g/mol) 411.034
Formel vægt 410.2991