Få mere at vide
Myosmine-2,4,5,6-d4, TRC
Artikelnummer. 30464319
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30464319 50 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30464319 Leverandør Toronto Research Chemicals Leverandørnr. TRCM83501050MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Myosmine-2,4,5,6-d4
CAS 66148-17-2
Renhedsgrad noter HPLC
Molekylær formel C9 2H4 H6 N2
Mængde 50 mg
Synonym Pyridine-2,3,4,6-d4, 5-(3,4-dihydro-2H-pyrrol-5-yl)-, Myosmine-2,4,5,6-d4, Myosmine-D4 (pyridine-D4)
InChI formel InChI=1S/C9H10N2/c1-3-8(7-10-5-1)9-4-2-6-11-9/h1,3,5,7H,2,4,6H2/i1D,3D,5D,7D
SMIL [2H]c1nc([2H])c(C2=NCCC2)c([2H])c1[2H]
IUPAC navn 2,3,4,6-tetradeuterio-5-(3,4-dihydro-2H-pyrrol-5-yl)pyridine
Molekylvægt (g/mol) 150.21
Formel vægt 150.1095
Procent renhed >95