Få mere at vide
Invitrogen™ Myosin 4 Monoclonal Antibody (MF20), Alexa Fluor™ 488, eBioscience™, Invitrogen™
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  Invitrogen™ 53-6503-82
 Se flere versioner af dette produkt
Produktkode 15302060
  • 3990.00 DKK / 100µg
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Denne vare kan ikke returneres. Se returpolitik
Beskrivelse
Beskrivelse
Description: This MF20 monoclonal antibody recognizes the heavy chain of myosin II, specificially the light meromyosin portion, in cardiac and skeletal muscle of vertebrates. Myosin II is composed of two heavy chains and four light chains. The 220-kDa myosin heavy chain exists as four different isoforms due to alternative splicing. Myosins interact with actin and hydrolyze ATP to function in muscle contraction, cytokinesis, and phagocytosis. The MF20 has been shown to react to myosin from a variety of mammalian, avian and amphibian species, including rat, mouse, human, chicken, zebrafish, and dog. Applications Reported: This MF20 antibody has been reported for use in immunohistochemical staining, immunocytochemistry, and immunohistochemical staining of frozen tissue sections. Applications Tested: This MF20 antibody has been tested by immunocytochemistry on fixed and permeabilized C2C12 cells that were differentiated for 2 days in low serum medium prior to staining. Staining can be performed using less than or equal to 5 μg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 488 nm; Emission: 519 nm; Laser: Blue Laser. Filtration: 0.2 μm post-manufacturing filtered.
Target protein includes a fast myosin isoform expressed in skeletal muscle and has function involved in Muscle contraction.
TRUSTED_SUSTAINABILITY
Tekniske data
Tekniske data
Myosin 4
Monoclonal
0.5 mg/mL
PBS with 0.09% sodium azide; pH 7.2
Q076A5, Q28641, Q29RW1, Q5SX39, Q9Y623
MYH4
Affinity chromatography
RUO
100009548, 100125991, 100726456, 17884, 360543, 408020, 4622, 468396, 479502
4°C, store in dark, DO NOT FREEZE!
Liquid
Responsibly packaged
Immunohistochemistry (Frozen), Western Blot, Immunocytochemistry
MF20
Alexa Fluor 488
MYH4
2B myosin heavy chain; AI506973; fast myosin heavy chain 2B; LOW QUALITY PROTEIN: myosin-4; MHC; MHC2B; Minimsc; Minmus; MM; MYH1; MYH2B; MYH-2B; myh4; MyHC IIb isoform; myHC-2b; MyHC-IIb; Myhsf; myosin heavy chain; myosin heavy chain 1; myosin heavy chain 2b; myosin heavy chain 2X; myosin heavy chain 4; Myosin heavy chain IIb; myosin heavy chain IIb isoform; myosin heavy chain isoform 2b; myosin heavy chain type IIb; Myosin heavy chain, skeletal muscle; myosin heavy chain, skeletal muscle, fetal; Myosin heavy chain, skeletal muscle, juvenile; myosin, heavy chain 4, skeletal muscle; myosin, heavy polypeptide 4, skeletal muscle; myosin-4; unnamed protein product
Mouse
100 μg
Primary
Bovine, Canine, Chicken, Chimpanzee, Feline, Guinea Pig, Human, Mouse, Monkey, Rabbit, Rat
Antibody
IgG2b κ
Suggestions de produits
Suggestions de produits
Vidéos
FDS
Documents
Documents
Product Certifications
Promotions
Promotions
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.
Merci de nous aider à améliorer notre site web. Votre commentaire a été soumis
For Research Use Only.