missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Myogenin Antibody (rMYOG/6297), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-14012AF532
Additional Details : Weight : 0.00970kg
Description
Myogenin Monoclonal antibody specifically detects Myogenin in Human samples. It is validated for Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
Myogenin | |
Monoclonal | |
Alexa Fluor 532 | |
BHLHC3, bHLHc3Myogenic factor-4; myogenin, Class C basic helix-loop-helix protein 3, myf-4, MYF4myogenin, Myogenic factor 4, MYOGENIN, myogenin (myogenic factor 4) | |
Recombinant full-length human Myogenin protein (Uniprot: P15173) | |
0.1 mL | |
Cancer, Stem Cell Markers, Transcription Factors and Regulators | |
4656 | |
Store at 4°C in the dark. | |
IgG1 κ |
Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
rMYOG/6297 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |